7-Methylxanthin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 7-Methyl-3,7-dihydro-1H-purin-2,6-dion, 3,7-Dihydro-7-methyl-1H-purin-2,6-dion, 7-Methyl-1H,3H-purin-2,6-dion, 2,6-Dihydroxy-7-methylpurin, 7-Methyl-3,7-dihydro-2H-purin-2,6-dion, 7-Methyl-2,6(1H,3H)-purindion

69723-250MG 69723-1G
SIAL69723-250MGEA 221 EUR
SIAL69723-250MG SIAL69723-1G
7-Methylxanthin, Sigma-Aldrich®
7-Methylxanthin
Formel: C₆H₆N₄O₂
Molecular Weight: 166,14 g/mol
MDL: MFCD00037979
CAS-Nummer: 552-62-5
EINECS: 209-019-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR