3-Isobutyl-1-methylxanthin 99+%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Isobutyl-1-methyl-2,6(1H,3H)-purindion, 3-Isobutyl-1-methyl-3,7-dihydro-1H-purin-2,6-dion, 3-Isobutyl-1-methyl-3,7-dihydro-2H-purin-2,6-dion, 3,7-Dihydro-3-isobutyl-1-methyl-1H-purin-2,6-dion, 3-Isobutyl-1-methyl-1H,3H-purin-2,6-dion, 2,6-Dihydroxy-1-methyl-3-isobutylpurin

228420010 228420050
ACRO228420010EA 157 EUR
ACRO228420010 ACRO228420050
3-Isobutyl-1-methylxanthin 99+%
3-Isobutyl-1-methylxanthin
Formel: C₁₀H₁₄N₄O₂
Molecular Weight: 222,25 g/mol
Schmelzpunkt: 200…203 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00005584
CAS-Nummer: 28822-58-4
EINECS: 249-259-3

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 200 to 203 °C
Titration with HClO4 ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR