Theobromin 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3,7-Dimethylxanthin, 3,7-Dimethyl-3,7-dihydro-2H-purin-2,6-dion, 3,7-Dimethyl-2,6(1H,3H)-purindion, 2,6-Dihydroxy-3,7-dimethylpurin, 3,7-Dimethyl-3,7-dihydro-1H-purin-2,6-dion, 3,7-Dimethyl-1H,3H-purin-2,6-dion, 3,7-Dihydro-3,7-dimethyl-1H-purin-2,6-dion

258821000 258822500 258820010
ACRO258821000EA 97 EUR
ACRO258821000 ACRO258822500 ACRO258820010
Theobromin 99%
Theobromin
Formel: C₇H₈N₄O₂
Molecular Weight: 180,17 g/mol
Schmelzpunkt: 290…295 °C
Dichte: 1,419 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00022830
CAS-Nummer: 83-67-0
EINECS: 201-494-2
Merck Index: 12,09418

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C)
Titration with NaOH ≥98.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR