Theobromin 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,7-Dimethylxanthin, 3,7-Dimethyl-3,7-dihydro-2H-purin-2,6-dion, 3,7-Dimethyl-2,6(1H,3H)-purindion, 2,6-Dihydroxy-3,7-dimethylpurin, 3,7-Dimethyl-3,7-dihydro-1H-purin-2,6-dion, 3,7-Dimethyl-1H,3H-purin-2,6-dion, 3,7-Dihydro-3,7-dimethyl-1H-purin-2,6-dion
A11861.18
A11861.22
A11861.30
A11861.18EA
59.7
EUR
A11861.18
A11861.22
A11861.30
Theobromin 99%
Theobromin
Purity: 99%
CAS No.: 83-67-0
Molecular Formula: C7H8N4O2
Formula Weight: 180.17
Melting Point: 345-350°
Beilstein Reference: 16464
Merck Reference: 14,9282
MDL No.: MFCD00022830
Phosphosdiesterase inhibitor
Formel:
C₇H₈N₄O₂ Molecular Weight: 180.17 g/mol Schmelzpunkt: 345…350 °C Dichte: 1.50 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022830 CAS-Nummer: 83-67-0 EINECS: 201-494-2 Merck Index: 12,09418 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...