6-Benzylaminopurin
Lieferant: FLUOROCHEM
Synonyme:
Benzyl(purin-6-yl)amin, 6-Benzyladenin, N6-Benzyladenin
023324-25G
023324-5G
FLUO023324-25GEA
39.9
EUR
FLUO023324-25G
FLUO023324-5G
6-Benzylaminopurin
6-Benzylaminopurin
Formel:
C₁₂H₁₁N₅ Molecular Weight: 225,25 g/mol Schmelzpunkt: 230…234 °C |
MDL:
MFCD00005572 CAS-Nummer: 1214-39-7 EINECS: 214-927-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...