6-Benzylaminopurin

Lieferant: Serva

Synonyme: Benzyl(purin-6-yl)amin, 6-Benzyladenin, N6-Benzyladenin

14812.01 14812.02
SERA14812.01EA 0 EUR
SERA14812.01 SERA14812.02
6-Benzylaminopurin
6-Benzylaminopurin
Formel: C₁₂H₁₁N₅
Molecular Weight: 225,25 g/mol
Schmelzpunkt: 230…234 °C
MDL: MFCD00005572
CAS-Nummer: 1214-39-7
EINECS: 214-927-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR