6-Benzylaminopurin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Benzyl(purin-6-yl)amin, 6-Benzyladenin, N6-Benzyladenin

M-Clarity™ Qualitätsstufe = MQ200

B3408-100MG B3408-1G B3408-25G B3408-500MG
SIALB3408-100MGEA 12.7 EUR
SIALB3408-100MG SIALB3408-1G SIALB3408-25G SIALB3408-500MG
6-Benzylaminopurin, Sigma-Aldrich®
6-Benzylaminopurin
Formel: C₁₂H₁₁N₅
Molecular Weight: 225,25 g/mol
Schmelzpunkt: 230…234 °C
MDL: MFCD00005572
CAS-Nummer: 1214-39-7
EINECS: 214-927-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR