2,3-Dichlorpyrazin

Lieferant: FLUOROCHEM
Achtung

002757-5G 002757-25G 002757-10G
FLUO002757-5GEA 0 EUR
FLUO002757-5G FLUO002757-25G FLUO002757-10G
2,3-Dichlorpyrazin
2,3-Dichloropyrazine
Formel: C₄H₂Cl₂N₂
MDL: MFCD00040964
CAS-Nummer: 4858-85-9
EINECS: 225-460-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR