2-Chlorpyrazin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Chloropyrazine
158060100
158060500
AC15806.10
AC15806.50
ACRO158060100EA
59.7
EUR
ACRO158060100
ACRO158060500
MAYBAC15806.10
MAYBAC15806.50
2-Chlorpyrazin 98%
2-Chlorpyrazin
Formel:
C₄H₃ClN₂ Molecular Weight: 114.53 g/mol Siedepunkt: 153 Dichte: 1.28 g/cm³ Flammpunkt: 57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006124 CAS-Nummer: 14508-49-7 EINECS: 238-517-0 UN: 1993 ADR: 3 III |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5320 to 1.5390 (20°C, 589 nm) |
Water | ≤3 % (K.F.) |
GC | ≥97.5 % |
UV | at 400 nm A: ≤0.01 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.0005 % |
Acidity | ≤0.0005 meq/g |
Heavy metals (as Pb) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...