3,5-Dimethyl-1H-pyrazol ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3,5-Dimethyl-1H-pyrazol

A10157.0B A10157.18 A10157.30
A10157.0BEA 324 EUR
A10157.0B A10157.18 A10157.30
3,5-Dimethyl-1H-pyrazol ≥99%
3,5-Dimethyl-1H-pyrazol

Purity: 99% CAS No.: 67-51-6 Molecular Formula: C5H8N2 Formula Weight: 96.13 pH: 6.7 Melting Point: 105-108° Boiling Point: 218° Beilstein Reference: 106325 MDL No.: MFCD00005243

Formel: C₅H₈N₂
Molecular Weight: 96,13 g/mol
Siedepunkt: 218 °C
Schmelzpunkt: 105…108 °C
Dichte: 0,8839 g/cm³ (16 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005243
CAS-Nummer: 67-51-6
EINECS: 200-657-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR