3,5-Dimethyl-1H-pyrazol

Lieferant: CHEMPUR

Synonyme: 3,5-Dimethyl-1H-pyrazol

SB01330.250 SB01330.1
CHMPSB01330.250EA 0 EUR
CHMPSB01330.250 MAYBSB01330.1
3,5-Dimethyl-1H-pyrazol
3,5-Dimethyl-1H-pyrazol
Formel: C₅H₈N₂
Molecular Weight: 96.13 g/mol
Siedepunkt: 218
Dichte: 0.8839 g/cm³ (16 °C)
MDL: MFCD00005243
CAS-Nummer: 67-51-6
EINECS: 200-657-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR