3-Amino-6-chlorpyridazin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Chlorpyridazin-3-amin
A10185.06
A10185.14
A10185.06EA
87
EUR
A10185.06
A10185.14
3-Amino-6-chlorpyridazin 98%
3-Amino-6-chlorpyridazin
Purity: 98%
CAS No.: 5469-69-2
Molecular Formula: C4H4ClN3
Formula Weight: 129.55
Melting Point: ca 218° dec.
Beilstein Reference: 110918
Formel:
C₄H₄ClN₃ Molecular Weight: 129,55 g/mol Schmelzpunkt: ∼ 218 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051506 CAS-Nummer: 5469-69-2 EINECS: 226-796-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...