3-Amino-6-chlorpyridazin

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Chlorpyridazin-3-amin

BTB05877.1 BTB05877EA BTB05877.50
MAYBBTB05877.1EA 0 EUR
MAYBBTB05877.1 MAYBBTB05877EA MAYBBTB05877.50
3-Amino-6-chlorpyridazin
3-Amino-6-chlorpyridazin
Formel: C₄H₄ClN₃
Molecular Weight: 129.55 g/mol
MDL: MFCD00051506
CAS-Nummer: 5469-69-2
EINECS: 226-796-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR