3-Amino-6-chlorpyridazin

Lieferant: FLUOROCHEM
Achtung

Synonyme: 6-Chlorpyridazin-3-amin

CFF759. 018176-1G 018176-5G 018176-25G
CHMPCFF759.EA 0 EUR
CHMPCFF759. FLUO018176-1G FLUO018176-5G FLUO018176-25G
3-Amino-6-chlorpyridazin
3-Amino-6-chlorpyridazin
Formel: C₄H₄ClN₃
Molecular Weight: 129.55 g/mol
MDL: MFCD00051506
CAS-Nummer: 5469-69-2
EINECS: 226-796-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR