3-Chlor-6-methylpyridazin

Lieferant: Thermo Scientific

BTB06842.250 BTB06842DA
MAYBBTB06842.250EA 0 EUR
MAYBBTB06842.250 MAYBBTB06842DA
3-Chlor-6-methylpyridazin
3-Chloro-6-methylpyridazine
Formel: C₅H₅ClN₂
Molecular Weight: 128,56 g/mol
MDL: MFCD00052905
CAS-Nummer: 1121-79-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR