2,3-Dibrompyridin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
705462-1G
705462-5G
SIAL705462-1GEA
26
EUR
SIAL705462-1G
SIAL705462-5G
2,3-Dibrompyridin, Sigma-Aldrich®
2,3-Dibromopyridine
Formel:
C₅H₃Br₂N Molecular Weight: 236,89 g/mol Siedepunkt: 249…249,5 °C (1013 hPa) Schmelzpunkt: 58…59 °C |
MDL:
MFCD00234014 CAS-Nummer: 13534-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...