2-Brom-3-chlorpyridin ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
L20012.03
L20012.06
L20012.03EA
22.7
EUR
L20012.03
L20012.06
2-Brom-3-chlorpyridin ≥97%
2-Bromo-3-chloropyridine
Purity: 97%
CAS No.: 96424-68-9
Molecular Formula: C5H3BrClN
Formula Weight: 192.44
Melting Point: 59-61°
Beilstein Reference: 1561638
Formel:
C₅H₃BrClN Schmelzpunkt: 59…61 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039344 CAS-Nummer: 96424-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...