2-Brom-4-chlorpyridin
Lieferant: FLUOROCHEM
012729
012729-1G
012729-5G
FLUO012729EA
0
EUR
FLUO012729
FLUO012729-1G
FLUO012729-5G
2-Brom-4-chlorpyridin
2-Bromo-4-chloropyridine
Formel:
C₅H₃BrClN Molecular Weight: 192.44 g/mol |
MDL:
MFCD03840757 CAS-Nummer: 22918-01-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...