3-Brom-4-chlorpyridin 98%

Lieferant: Thermo Fisher Scientific
Achtung

379640010 379640050
ACRO379640010EA 74 EUR
ACRO379640010 ACRO379640050
3-Brom-4-chlorpyridin 98%
3-Bromo-4-chloropyridine
Formel: C₅H₃BrClN
Molecular Weight: 192.44 g/mol
MDL: MFCD00234008
CAS-Nummer: 36953-42-1

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR