4-Brom-2-chlorpyridin, Sigma-Aldrich®

Lieferant: Merck

646318-25G 646318-5G
SIAL646318-25GEA 140 EUR
SIAL646318-25G SIAL646318-5G
4-Brom-2-chlorpyridin, Sigma-Aldrich®
4-Bromo-2-chloropyridine
Formel: C₅H₃BrClN
Molecular Weight: 192,44 g/mol
MDL: MFCD03840756
CAS-Nummer: 73583-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR