5-Brom-2-chlornicotinsäure

Lieferant: BIONET RESEARCH

10W-0709.500 10W-0709.1 10W-0709.5 10W-0709.10
BIOT10W-0709.500EA 0 EUR
BIOT10W-0709.500 BIOT10W-0709.1 BIOT10W-0709.5 BIOT10W-0709.10
5-Brom-2-chlornicotinsäure
5-Brom-2-chlornicotinsäure
Formel: C₆H₃BrClNO₂
MDL: MFCD03844847
CAS-Nummer: 29241-65-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR