2-Amino-3-brom-5-chlorpyridin
Lieferant: BIONET RESEARCH
|
Achtung
|
Synonyme:
3-Brom-5-chlorpyridin-2-amin, 3-Brom-5-chlor-2-pyridylamin
4T-0646.500
4T-0646.1
4T-0646.5
4T-0646.10
KO4T-0646.0001
BIOT4T-0646.500EA
0
EUR
BIOT4T-0646.500
BIOT4T-0646.1
BIOT4T-0646.5
BIOT4T-0646.10
ABCRKO4T-0646.0001
2-Amino-3-brom-5-chlorpyridin
2-Amino-3-brom-5-chlorpyridin
Formel:
C₅H₄BrClN₂ Molecular Weight: 207,46 g/mol |
MDL:
MFCD00955627 CAS-Nummer: 26163-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...