6-Bromnicotinsäure

Lieferant: FLUOROCHEM

Synonyme: 6-Brom-3-pyridincarbonsäure, 6-Brompyridin-3-carbonsäure

023530-25G 023530-5G
FLUO023530-25GEA 112 EUR
FLUO023530-25G FLUO023530-5G
6-Bromnicotinsäure
6-Bromnicotinsäure
Formel: C₆H₄BrNO₂
Molecular Weight: 202,01 g/mol
MDL: MFCD01927100
CAS-Nummer: 6311-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR