2-Amino-5-bromnicotinsäure

Lieferant: BIONET RESEARCH

11W-0705.500 11W-0705.1 11W-0705.5 11W-0705.10
BIOT11W-0705.500EA 0 EUR
BIOT11W-0705.500 BIOT11W-0705.1 BIOT11W-0705.5 BIOT11W-0705.10
2-Amino-5-bromnicotinsäure
2-Amino-5-bromnicotinsäure
Formel: C₆H₅BrN₂O₂
Molecular Weight: 217.02 g/mol
MDL: MFCD01860227
CAS-Nummer: 52833-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR