2,4,6-Tribrom-3,5-difluorpyridin
Lieferant: FLUOROCHEM
Synonyme:
3,5-Difluoro-2,4,6-tribromopyridine
010342-1G
010342-2.5G
FLUO010342-1GEA
0
EUR
FLUO010342-1G
FLUO010342-2.5G
2,4,6-Tribrom-3,5-difluorpyridin
2,4,6-Tribrom-3,5-difluorpyridin
Formel:
C₅Br₃F₂N Molecular Weight: 351,77 g/mol |
CAS-Nummer:
30841-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...