5-Brom-2-iodpyridin

Lieferant: FLUOROCHEM
Achtung

023885-25G 023885-5G
FLUO023885-25GEA 76.2 EUR
FLUO023885-25G FLUO023885-5G
5-Brom-2-iodpyridin
5-Brom-2-iodpyridin
Formel: C₅H₃BrIN
Molecular Weight: 283,89 g/mol
MDL: MFCD00691506
CAS-Nummer: 223463-13-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR