5-Brom-2-iodpyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung

569607-25G 569607-5G
SIAL569607-25GEA 88.4 EUR
SIAL569607-25G SIAL569607-5G
5-Brom-2-iodpyridin, Sigma-Aldrich®
5-Brom-2-iodpyridin
Formel: C₅H₃BrIN
Molecular Weight: 283,89 g/mol
MDL: MFCD00691506
CAS-Nummer: 223463-13-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR