2-Amino-3,5-dibrompyridin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3,5-Dibrompyridin-2-amin, 3,5-Dibrom-2-pyridylamin
A12706.14
A12706.22
A12706.14EA
94.3
EUR
A12706.14
A12706.22
2-Amino-3,5-dibrompyridin 97%
2-Amino-3,5-dibrompyridin
Purity: 97%
CAS No.: 35486-42-1
Molecular Formula: C5H4Br2N2
Formula Weight: 251.92
Melting Point: 102-105°
Beilstein Reference: 119390
Formel:
C₅H₄Br₂N₂ Molecular Weight: 251,91 g/mol Siedepunkt: 254 °C (1013 hPa) Schmelzpunkt: 102…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038041 CAS-Nummer: 35486-42-1 EINECS: 252-590-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...