2-Amino-3-brompyridin 98%

Lieferant: Thermo Scientific
Achtung

360580050 360580250
ACRO360580050EA 46 EUR
ACRO360580050 ACRO360580250
2-Amino-3-brompyridin 98%
2-Amino-3-brompyridin
Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
Schmelzpunkt: 61…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095187
CAS-Nummer: 13534-99-1

Bestellen

Spezifikation Testergebnisse

Appearance Grey to brown Powder
Infrared spectrum Conforms
Melting point 61 to 65 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR