2-Amino-3-brompyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

553719-25G 553719-5G
SIAL553719-25GEA 156 EUR
SIAL553719-25G SIAL553719-5G
2-Amino-3-brompyridin, Sigma-Aldrich®
2-Amino-3-brompyridin
Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
MDL: MFCD03095187
CAS-Nummer: 13534-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR