2-Amino-5-brompyridin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Brompyridin-2-amin, 5-Brom-2-pyridylamin
153740050
153740250
ACRO153740050EA
31.1
EUR
ACRO153740050
ACRO153740250
2-Amino-5-brompyridin 97%
2-Amino-5-brompyridin
Formel:
C₅H₅BrN₂ Molecular Weight: 173,01 g/mol Siedepunkt: 244 °C (1013 hPa) Schmelzpunkt: 135…138 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006323 CAS-Nummer: 1072-97-5 EINECS: 214-019-9 |
Spezifikation Testergebnisse
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 135 to 140 °C |
GC | ≥96.0 % |
Residue after evaporation | ≤0.002 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...