2-Amino-5-brompyridin 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Brompyridin-2-amin, 5-Brom-2-pyridylamin

A14280.06 A14280.14 A14280.22
A14280.06EA 30.6 EUR
A14280.06 A14280.14 A14280.22
2-Amino-5-brompyridin 98%
2-Amino-5-brompyridin

Purity: 98% CAS No.: 1072-97-5 Molecular Formula: C5H5BrN2 Formula Weight: 173.01 Melting Point: 135-140° Beilstein Reference: 108737

Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
Siedepunkt: 244 °C (1013 hPa)
Schmelzpunkt: 135…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006323
CAS-Nummer: 1072-97-5
EINECS: 214-019-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR