2-Amino-6-brompyridin 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-Brompyridin-2-amin, 6-Brom-2-pyridylamin

H26555.06 H26555.14
H26555.06EA 85.8 EUR
H26555.06 H26555.14
2-Amino-6-brompyridin 98%
2-Amino-6-brompyridin

Purity: 98% CAS No.: 19798-81-3 Molecular Formula: C5H5BrN2 Formula Weight: 173.01 Melting Point: 88-91° MDL No.: MFCD00137843

Formel: C₅H₅BrN₂
Molecular Weight: 173.01 g/mol
Schmelzpunkt: 88…91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00137843
CAS-Nummer: 19798-81-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR