3-Amino-5-brompyridin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
5-Brom-3-pyridylamin, 5-Brompyridin-3-amin
M-Clarity™ Qualitätsstufe = MQ100
667129-10G
667129-1G
SIAL667129-10GEA
43.2
EUR
SIAL667129-10G
SIAL667129-1G
3-Amino-5-brompyridin, Sigma-Aldrich®
3-Amino-5-brompyridin
Formel:
C₅H₅BrN₂ Molecular Weight: 173,01 g/mol |
MDL:
MFCD01646060 CAS-Nummer: 13535-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...