5-Amino-2-brompyridin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Brompyridin-5-amin, 2-Brom-5-pyridylamin

L20008.06 L20008.14
L20008.06EA 111 EUR
L20008.06 L20008.14
5-Amino-2-brompyridin 97%
5-Amino-2-brompyridin

Purity: 97% CAS No.: 13534-97-9 Molecular Formula: C5H5BrN2 Formula Weight: 173.01 Melting Point: 75-79° Beilstein Reference: 109102

Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
Schmelzpunkt: 75…79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01646116
CAS-Nummer: 13534-97-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR