4-Amino-2-brompyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Brom-4-pyridylamin, 2-Brompyridin-4-amin

M-Clarity™ Qualitätsstufe = MQ200

654086-25G
SIAL654086-25GEA 335 EUR
SIAL654086-25G
4-Amino-2-brompyridin, Sigma-Aldrich®
4-Amino-2-brompyridin
Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
MDL: MFCD01646061
CAS-Nummer: 7598-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR