4-Amino-3-brompyridin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Brompyridin-4-amin, 3-Brom-4-pyridylamin
AC36062.1
AC36062.10
360620010
360620050
MAYBAC36062.1EA
0
EUR
MAYBAC36062.1
MAYBAC36062.10
ACRO360620010
ACRO360620050
4-Amino-3-brompyridin 98%
4-Amino-3-brompyridin
Formel:
C₅H₅BrN₂ Molecular Weight: 173,01 g/mol Siedepunkt: 244 °C (1013 hPa) Schmelzpunkt: 67…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02068297 CAS-Nummer: 13534-98-0 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 67 to 71 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...