2-Brom-5-nitropyridin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
324833-1G
324833-5G
SIAL324833-1GEA
25.9
EUR
SIAL324833-1G
SIAL324833-5G
2-Brom-5-nitropyridin, Sigma-Aldrich®
2-Bromo-5-nitropyridine
Formel:
C₅H₃BrN₂O₂ Molecular Weight: 203 g/mol |
MDL:
MFCD00006222 CAS-Nummer: 4487-59-6 EINECS: 224-777-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...