3-Brom-5-methoxypyridin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
5-Bromo-3-methoxypyridine
M-Clarity™ Qualitätsstufe = MQ100
631817-1G
631817-5G
SIAL631817-1GEA
27.3
EUR
SIAL631817-1G
SIAL631817-5G
3-Brom-5-methoxypyridin, Sigma-Aldrich®
3-Bromo-5-methoxypyridine
Formel:
C₆H₆BrNO Molecular Weight: 188,02 g/mol |
MDL:
MFCD00234169 CAS-Nummer: 50720-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...