5-Brom-2-methoxypyridin
Lieferant: FLUOROCHEM
|
Achtung
|
023910-25G
023910-5G
FLUO023910-25GEA
45.9
EUR
FLUO023910-25G
FLUO023910-5G
5-Brom-2-methoxypyridin
5-Brom-2-methoxypyridin
Formel:
C₆H₆BrNO Molecular Weight: 188,02 g/mol |
MDL:
MFCD01318952 CAS-Nummer: 13472-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...