5-Brom-2-methoxypyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ200

510297-25ML 510297-5ML
SIAL510297-25MLEA 192 EUR
SIAL510297-25ML SIAL510297-5ML
5-Brom-2-methoxypyridin, Sigma-Aldrich®
5-Brom-2-methoxypyridin
Formel: C₆H₆BrNO
Molecular Weight: 188,02 g/mol
MDL: MFCD01318952
CAS-Nummer: 13472-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR