2-Brom-3-hydroxypyridin 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Brom-3-pyridinol
152170050
152170250
ACRO152170050EA
38.4
EUR
ACRO152170050
ACRO152170250
2-Brom-3-hydroxypyridin 99%
2-Brom-3-hydroxypyridin
Formel:
C₅H₄BrNO Molecular Weight: 174 g/mol Siedepunkt: 232 Schmelzpunkt: 185…188 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006220 CAS-Nummer: 6602-32-0 EINECS: 229-547-5 |
Spezifikation Testergebnisse
Appearance | Grey to light brown Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...