2-Brom-5-hydroxypyridin 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Brom-5-pyridinol

H27228.06 H27228.03
H27228.06EA 314 EUR
H27228.06 H27228.03
2-Brom-5-hydroxypyridin 95%
2-Brom-5-hydroxypyridin

Purity: 95% CAS No.: 55717-45-8 Molecular Formula: C5H4BrNO Formula Weight: 174.00 Melting Point: 133-136° MDL No.: MFCD04114184

Formel: C₅H₄BrNO
Molecular Weight: 174 g/mol
Schmelzpunkt: 133…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114184
CAS-Nummer: 55717-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR