5-Brom-2-hydroxypyridin ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Brom-6-pyridinol, 3-Brom-6-hydroxypyridin, 5-Brom-2-pyridinol
B25267.06
B25267.14
B25267.06EA
24.4
EUR
B25267.06
B25267.14
5-Brom-2-hydroxypyridin ≥97%
5-Brom-2-hydroxypyridin
Purity: 97%
CAS No.: 13466-38-1
Molecular Formula: C5H4BrNO
Formula Weight: 174.00
Melting Point: 178-182°
Beilstein Reference: 108751
Formel:
C₅H₄BrNO Molecular Weight: 174 g/mol Schmelzpunkt: 178…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234042 CAS-Nummer: 13466-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...