2,3,5-Trichlorpyridin
Lieferant: CHEMPUR
Synonyme:
2,3,5-TCP
BTB14380.250
BTB14380.1
CHMPBTB14380.250EA
0
EUR
CHMPBTB14380.250
MAYBBTB14380.1
2,3,5-Trichlorpyridin
2,3,5-Trichlorpyridin
Formel:
C₅H₂Cl₃N Molecular Weight: 182.44 g/mol |
MDL:
MFCD00043007 CAS-Nummer: 16063-70-0 EINECS: 407-270-2 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...