2,4,6-Trichlorpyridin

Lieferant: Thermo Scientific

BTB15034.250 BTB15034.1
MAYBBTB15034.250EA 0 EUR
MAYBBTB15034.250 MAYBBTB15034.1
2,4,6-Trichlorpyridin
2,4,6-Trichlorpyridin
Formel: C₅H₂Cl₃N
Molecular Weight: 182.44 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00971928
CAS-Nummer: 16063-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR