2,4,6-Trichlorpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

633534-1G 633534-5G
SIAL633534-1GEA 82.2 EUR
SIAL633534-1G SIAL633534-5G
2,4,6-Trichlorpyridin, Sigma-Aldrich®
2,4,6-Trichlorpyridin
Formel: C₅H₂Cl₃N
Molecular Weight: 182.44 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00971928
CAS-Nummer: 16063-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR