2-Chlornicotinamid

Lieferant: Thermo Scientific
Achtung

BTB06407.1 BTB06407.10 BTB06407.50
MAYBBTB06407.1EA 0 EUR
MAYBBTB06407.1 MAYBBTB06407.10 MAYBBTB06407.50
2-Chlornicotinamid
2-Chlornicotinamid
Formel: C₆H₅ClN₂O
Molecular Weight: 156.57 g/mol
Schmelzpunkt: 164…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006237
CAS-Nummer: 10366-35-5
EINECS: 233-808-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR