2,5-Dichlornicotinsäure
Lieferant: BIONET RESEARCH
7W-0399.500
7W-0399.1
7W-0399.5
7W-0399.10
BIOT7W-0399.500EA
0
EUR
BIOT7W-0399.500
BIOT7W-0399.1
BIOT7W-0399.5
BIOT7W-0399.10
2,5-Dichlornicotinsäure
2,5-Dichlornicotinsäure
Formel:
C₆H₃Cl₂NO₂ Molecular Weight: 192 g/mol |
MDL:
MFCD01861979 CAS-Nummer: 59782-85-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...