2,6-Dichlornicotinsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,6-Dichlor-3-pyridincarbonsäure, 2,6-Dichlorpyridin-3-carbonsäure

L20002.03 L20002.06
L20002.03EA 35.5 EUR
L20002.03 L20002.06
2,6-Dichlornicotinsäure ≥97%
2,6-Dichlornicotinsäure

Purity: 97% CAS No.: 38496-18-3 Molecular Formula: C6H3Cl2NO2 Formula Weight: 192.00 Melting Point: 146-151° Beilstein Reference: 136114

Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192 g/mol
Schmelzpunkt: 146…151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075583
CAS-Nummer: 38496-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR